Assay Details
| Gene Symbol: Lrrc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019457.2 | NP_062330.1 | 8 - 9 | 1114 | 122 | |
| GenBank mRNA | AF092208.1 | - | 8 - 9 | 1059 | 122 | |
| AK077124.1 | - | 8 - 9 | 1098 | 122 | ||
| BC046277.1 | - | 8 - 9 | 1065 | 6310345 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc6
Genomic Map