Assay Details
| Gene Symbol: Jak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048177.2 | NP_001041642.1 | 15 - 16 | 2470 | 75 | |
| NM_008413.3 | NP_032439.2 | 16 - 17 | 2553 | 75 | ||
| GenBank mRNA | BC054807.1 | - | 15 - 16 | 2233 | 6838318 | 75 |
| BC059834.1 | - | 15 - 16 | 2233 | 6838318 | 75 | |
| L16956.1 | - | 15 - 16 | 2222 | 75 | ||
More Information
Set Membership: |
|

Jak2
Genomic Map