Assay Details
| Gene Symbol: Plgrkt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026362.2 | NP_080638.1 | 3 - 4 | 270 | 142 | |
| GenBank mRNA | AK017215.1 | - | 3 - 4 | 271 | 142 | |
| AK168454.1 | - | 3 - 4 | 246 | 142 | ||
| AK170700.1 | - | 4 - 5 | 344 | 142 | ||
| BC024953.1 | - | 3 - 4 | 229 | 4951101 | 142 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK017215.1, AK168454.1, AK170700.1, BC024953.1, NM_026362.2 transcripts.
Set Membership: |
|

Plgrkt
Genomic Map