Assay Details
| Gene Symbol: Dydc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027094.1 | NP_081370.1 | 2 - 3 | 295 | 120 | |
| GenBank mRNA | AK006511.1 | - | 2 - 3 | 295 | 120 | |
| BC049632.1 | - | 2 - 3 | 294 | 6771945 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dydc1
Genomic Map