Assay Details
| Gene Symbol: Dydc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027094.1 | NP_081370.1 | 7 - 8 | 787 | 86 | |
| GenBank mRNA | AK006511.1 | - | 7 - 8 | 787 | 86 | |
| BC049632.1 | - | 7 - 8 | 738 | 6771945 | 86 | |
| BX632188.1 | - | 1 - 2 | 86 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P388471 | AK006511.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P388471 | BC049632.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P388471 | BX632188.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P388471 | NM_027094.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P388471 | AK006511.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P388471 | BC049632.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P388471 | BX632188.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P388471 | NM_027094.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 538748 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 173742 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 566802 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 110124 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 93147 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 572070 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 795454 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 291278 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 516274 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 895291 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 766947 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 951264 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 708692 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 111082 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 40520 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105504 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107192 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154133 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P388471 | AK006511.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P388471 | BC049632.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P388471 | BX632188.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P388471 | NM_027094.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P388471 | AK006511.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P388471 | BC049632.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P388471 | BX632188.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P388471 | NM_027094.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P105504 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P107192 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08093418 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154133 | - | - |
| Whole Mouse Genome G4122A | A_51_P388471 | AK006511.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P388471 | BC049632.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P388471 | BX632188.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P388471 | NM_027094.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P388471 | AK006511.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P388471 | BC049632.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P388471 | BX632188.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P388471 | NM_027094.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P107192 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154133 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P105504 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29561133 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11665775 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29561131 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29561138 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29561130 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29561140 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10414183 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10414189 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10414184 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10414187 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10414190 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10414188 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10414185 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10414191 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453205_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10414182 | - | - |
| Mouse Genome 430 2.0 Array | 1453205_at | - | - |
| Mouse Genome 430 2.0 Array | 1453205_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69496_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69496_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400003567.mm.1 | NM_027094.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400003567.mm.1 | NM_027094.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400003567.mm.1 | TC1400000410.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400003566.mm.1 | NM_027094.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400003566.mm.1 | NM_027094.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400003566.mm.1 | TC1400000410.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1453205_at | - | - |
| Mouse Expression Set 430 | 1453205_PM_at | - | - |
| Murine Genome U74v2 Set | 166057_r_at | - | - |
| Murine Genome U74v2 Set | 166057_PM_r_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453205_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453205_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 842993 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1226008 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1226008 | - | - |

Dydc1
Genomic Map