Assay Details
| Gene Symbol: Usp9y | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148943.2 | NP_683745.2 | 31 - 32 | 4798 | 139 | |
| GenBank mRNA | AJ307017.1 | - | 31 - 32 | 4798 | 139 | |
More Information
Set Membership: |
|

Usp9y
Genomic Map