Assay Details
| Gene Symbol: Usp9y | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148943.2 | NP_683745.2 | 9 - 10 | 1191 | 138 | |
| GenBank mRNA | AJ307017.1 | - | 9 - 10 | 138 | ||
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0Y00000010.rn
Set Membership: |
|

Usp9y
Genomic Map