Assay Details
| Gene Symbol: Gnpda2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038015.1 | NP_001033104.1 | 1 - 2 | 82 | 57 | |
| GenBank mRNA | AK014950.1 | - | 1 - 2 | 92 | 57 | |
| AK015527.1 | - | 1 - 2 | 92 | 57 | ||
| AK017588.1 | - | 1 - 2 | 91 | 57 | ||
| AK051596.1 | - | 1 - 2 | 91 | 57 | ||
| AK141408.1 | - | 1 - 2 | 94 | 57 | ||
| AK166601.1 | - | 1 - 2 | 84 | 57 | ||
| AK168516.1 | - | 1 - 2 | 101 | 57 | ||
| AK171299.1 | - | 1 - 2 | 77 | 57 | ||
| AK220275.1 | - | 1 - 2 | 43 | 57 | ||
| BC004084.1 | - | 1 - 2 | 82 | 3593102 | 57 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK015527.1, AK017588.1, AK141408.1, AK166601.1, AK168516.1, AK171299.1, BC004084.1, NM_001038015.1 transcripts.
Set Membership: |
|

Gnpda2
Genomic Map