Assay Details
| Gene Symbol: Nsun7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014968.1 | - | 2 - 3 | 149 | 144 | |
| AK035181.1 | - | 3 - 4 | 664 | 144 | ||
| BC115878.1 | - | 2 - 3 | 473 | 40088737 | 144 | |
| BC116658.1 | - | 2 - 3 | 473 | 40088738 | 144 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK014968.1 transcripts.
Set Membership: |
|

Nsun7
Genomic Map