Assay Details
| Gene Symbol: Nsun7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014968.1 | - | 3 - 4 | 281 | 109 | |
| AK035181.1 | - | 4 - 5 | 796 | 109 | ||
| BC115878.1 | - | 3 - 4 | 605 | 40088737 | 109 | |
| BC116658.1 | - | 3 - 4 | 605 | 40088738 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nsun7
Genomic Map