Assay Details
| Gene Symbol: Ppargc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008904.2 | NP_032930.1 | 8 - 9 | 1923 | 88 | |
| NR_027710.1 | 8 - 9 | 1954 | 88 | |||
| GenBank mRNA | AB061324.1 | - | 8 - 9 | 1877 | 88 | |
| AF049330.1 | - | 8 - 9 | 1875 | 88 | ||
| AK138668.1 | - | 8 - 9 | 1924 | 88 | ||
| AK143753.1 | - | 8 - 9 | 1875 | 88 | ||
| JX866948.1 | - | 8 - 9 | 1842 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppargc1a
Genomic Map