Assay Details
| Gene Symbol: N4bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024917.1 | NP_001020088.1 | 8 - 9 | 1919 | 67 | |
| GenBank mRNA | AK048616.1 | - | 3 - 4 | 1175 | 67 | |
| AK173167.1 | - | 6 - 7 | 1630 | 67 | ||
| BC054549.1 | - | 8 - 9 | 1919 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

N4bp2
Genomic Map