Assay Details
| Gene Symbol: Ugdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009466.2 | NP_033492.1 | 2 - 3 | 385 | 79 | |
| GenBank mRNA | AF061017.1 | - | 2 - 3 | 269 | 79 | |
| AK144673.1 | - | 2 - 3 | 421 | 79 | ||
| AK146693.1 | - | 2 - 3 | 269 | 79 | ||
| AK162298.1 | - | 2 - 3 | 273 | 79 | ||
| AK167462.1 | - | 2 - 3 | 265 | 79 | ||
| AK167560.1 | - | 2 - 3 | 365 | 79 | ||
| AK168508.1 | - | 2 - 3 | 272 | 79 | ||
| AK168563.1 | - | 2 - 3 | 269 | 79 | ||
| AK169208.1 | - | 2 - 3 | 400 | 79 | ||
| BC006749.1 | - | 2 - 3 | 385 | 3708709 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ugdh
Genomic Map