Assay Details
| Gene Symbol: Klhl32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163020.1 | NP_001156492.1 | 5 - 6 | 821 | 87 | |
| GenBank mRNA | AK122569.1 | - | 444 | 87 | ||
| BC141239.1 | - | 4 - 5 | 426 | 9055851 | 87 | |
| BC145566.1 | - | 3 - 4 | 322 | 87 | ||
| CA556185.2 | - | 4 - 5 | 413 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Klhl32
Genomic Map