Assay Details
| Gene Symbol: Klhl32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033531.3 | NP_001028703.2 | 5 - 6 | 934 | 112 | |
| NM_001163020.1 | NP_001156492.1 | 6 - 7 | 1033 | 112 | ||
| GenBank mRNA | AK134944.1 | - | 5 - 6 | 906 | 112 | |
| BC141239.1 | - | 5 - 6 | 638 | 9055851 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Klhl32
Genomic Map