Assay Details
| Gene Symbol: Klhl32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033531.3 | NP_001028703.2 | 8 - 9 | 1913 | 59 | |
| NM_001163020.1 | NP_001156492.1 | 9 - 10 | 2012 | 59 | ||
| GenBank mRNA | AK122569.1 | - | 1629 | 59 | ||
| AK134944.1 | - | 8 - 9 | 1885 | 59 | ||
| BC141239.1 | - | 8 - 9 | 1617 | 9055851 | 59 | |
| BC145566.1 | - | 7 - 8 | 1509 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Klhl32
Genomic Map