Assay Details
| Gene Symbol: Map9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081230.1 | NP_001074699.1 | 10 - 11 | 1632 | 150 | |
| GenBank mRNA | AK030527.1 | - | 10 - 11 | 1473 | 150 | |
| AK135702.1 | - | 7 - 8 | 1126 | 150 | ||
| AK135823.1 | - | 10 - 11 | 1536 | 150 | ||
| AK138540.1 | - | 10 - 11 | 1577 | 150 | ||
| AK147287.1 | - | 10 - 11 | 1575 | 150 | ||
| AK147416.1 | - | 10 - 11 | 1575 | 150 | ||
| BC051460.1 | - | 2 - 3 | 157 | 150 | ||
| BC061216.1 | - | 10 - 11 | 1488 | 150 | ||
| BC087935.1 | - | 10 - 11 | 1529 | 150 | ||
More Information
Set Membership: |
|

Map9
Genomic Map