| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300004189.mm.1 |
NM_001081230.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300004189.mm.1 |
NM_001081230.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300004189.mm.1 |
TC0300000637.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300004191.mm.1 |
NM_001081230.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300004191.mm.1 |
NM_001081230.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300004191.mm.1 |
TC0300000637.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300004185.mm.1 |
NM_001081230.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300004185.mm.1 |
NM_001081230.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300004185.mm.1 |
TC0300000637.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300004187.mm.1 |
NM_001081230.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300004187.mm.1 |
NM_001081230.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300004187.mm.1 |
TC0300000637.mm.1 |
MTA TC |