Assay Details
| Gene Symbol: Map9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081230.1 | NP_001074699.1 | 5 - 6 | 965 | 95 | |
| GenBank mRNA | AK030527.1 | - | 5 - 6 | 806 | 95 | |
| AK038737.1 | - | 5 - 6 | 910 | 95 | ||
| AK135702.1 | - | 2 - 3 | 459 | 95 | ||
| AK135823.1 | - | 5 - 6 | 869 | 95 | ||
| AK138540.1 | - | 5 - 6 | 910 | 95 | ||
| AK147287.1 | - | 5 - 6 | 908 | 95 | ||
| AK147416.1 | - | 5 - 6 | 908 | 95 | ||
| AK162555.1 | - | 5 - 6 | 885 | 95 | ||
| BC061216.1 | - | 5 - 6 | 821 | 95 | ||
| BC087935.1 | - | 5 - 6 | 862 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map9
Genomic Map