Assay Details
| Gene Symbol: Map9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081230.1 | NP_001074699.1 | 7 - 8 | 1305 | 110 | |
| GenBank mRNA | AK030527.1 | - | 7 - 8 | 1146 | 110 | |
| AK135702.1 | - | 4 - 5 | 799 | 110 | ||
| AK135823.1 | - | 7 - 8 | 1209 | 110 | ||
| AK138540.1 | - | 7 - 8 | 1250 | 110 | ||
| AK147287.1 | - | 7 - 8 | 1248 | 110 | ||
| AK147416.1 | - | 7 - 8 | 1248 | 110 | ||
| AK162555.1 | - | 7 - 8 | 1225 | 110 | ||
| BC061216.1 | - | 7 - 8 | 1161 | 110 | ||
| BC087935.1 | - | 7 - 8 | 1202 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map9
Genomic Map