Assay Details
| Gene Symbol: Fbxw7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177773.1 | NP_001171244.1 | 10 - 11 | 1619 | 91 | |
| NM_001177774.1 | NP_001171245.1 | 9 - 10 | 1528 | 91 | ||
| NM_080428.3 | NP_536353.2 | 7 - 8 | 1164 | 91 | ||
| GenBank mRNA | AB221593.1 | - | 7 - 8 | 1006 | 91 | |
| AF391192.1 | - | 7 - 8 | 1164 | 91 | ||
| AK032174.1 | - | 1 - 2 | 90 | 91 | ||
| BC060620.1 | - | 10 - 11 | 1619 | 91 | ||
| BC066093.1 | - | 9 - 10 | 1528 | 91 | ||
| BC085184.1 | - | 9 - 10 | 3320 | 30664813 | 91 | |
| BC131648.1 | - | 7 - 8 | 1028 | 40110349 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxw7
Genomic Map