Assay Details
| Gene Symbol: Fbxw7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177773.1 | NP_001171244.1 | 12 - 13 | 2031 | 98 | |
| NM_001177774.1 | NP_001171245.1 | 11 - 12 | 1940 | 98 | ||
| NM_080428.3 | NP_536353.2 | 9 - 10 | 1576 | 98 | ||
| GenBank mRNA | AB221593.1 | - | 9 - 10 | 1418 | 98 | |
| AF391192.1 | - | 9 - 10 | 1576 | 98 | ||
| AF427101.1 | - | 9 - 10 | 1498 | 98 | ||
| AK032174.1 | - | 3 - 4 | 502 | 98 | ||
| BC060620.1 | - | 12 - 13 | 2031 | 98 | ||
| BC066093.1 | - | 11 - 12 | 1941 | 98 | ||
| BC085184.1 | - | 11 - 12 | 3732 | 30664813 | 98 | |
| BC131648.1 | - | 9 - 10 | 1440 | 40110349 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxw7
Genomic Map