Assay Details
| Gene Symbol: Fbxw7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177773.1 | NP_001171244.1 | 1 - 2 | 196 | 193 | |
| GenBank mRNA | BC060620.1 | - | 1 - 2 | 196 | 193 | |
| BC085184.1 | - | 1 - 2 | 295 | 30664813 | 193 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC085184.1, NM_001177773.1 transcripts.
Set Membership: |
|

Fbxw7
Genomic Map