Assay Details
| Gene Symbol: Fbxw7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177773.1 | NP_001171244.1 | 7 - 8 | 1249 | 74 | |
| NM_001177774.1 | NP_001171245.1 | 6 - 7 | 1158 | 74 | ||
| NM_080428.3 | NP_536353.2 | 4 - 5 | 794 | 74 | ||
| GenBank mRNA | AB221593.1 | - | 4 - 5 | 636 | 74 | |
| AF391192.1 | - | 4 - 5 | 794 | 74 | ||
| AF427101.1 | - | 4 - 5 | 716 | 74 | ||
| BC060620.1 | - | 7 - 8 | 1249 | 74 | ||
| BC066093.1 | - | 6 - 7 | 1158 | 74 | ||
| BC085184.1 | - | 7 - 8 | 1348 | 30664813 | 74 | |
| BC131648.1 | - | 4 - 5 | 658 | 40110349 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxw7
Genomic Map