Assay Details
| Gene Symbol: Tiparp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178892.5 | NP_849223.2 | 1 - 2 | 182 | 90 | |
| GenBank mRNA | AK028529.1 | - | 1 - 2 | 183 | 90 | |
| AK032453.1 | - | 1 - 2 | 166 | 90 | ||
| AK150248.1 | - | 1 - 2 | 164 | 90 | ||
| AK162094.1 | - | 1 - 2 | 188 | 90 | ||
| AK163347.1 | - | 1 - 2 | 221 | 90 | ||
| BC068173.1 | - | 1 - 2 | 156 | 30616032 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK028529.1, AK032453.1, AK150248.1, AK162094.1, BC068173.1, NM_178892.5 transcripts.
Set Membership: |
|

Tiparp
Genomic Map