Assay Details
| Gene Symbol: Hgsnat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029884.1 | NP_084160.1 | 9 - 10 | 944 | 102 | |
| GenBank mRNA | AK035264.1 | - | 9 - 10 | 877 | 102 | |
| AK149883.1 | - | 9 - 10 | 948 | 102 | ||
| AK152926.1 | - | 9 - 10 | 909 | 102 | ||
| AK159649.1 | - | 9 - 10 | 873 | 102 | ||
| AK160068.1 | - | 9 - 10 | 867 | 102 | ||
| AK209791.1 | - | 3 - 4 | 210 | 102 | ||
| BC024084.1 | - | 9 - 10 | 851 | 5101064 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hgsnat
Genomic Map