Assay Details
| Gene Symbol: Vstm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178791.4 | NP_848906.1 | 3 - 4 | 681 | 91 | |
| GenBank mRNA | AK033244.1 | - | 3 - 4 | 637 | 91 | |
| AK040699.1 | - | 3 - 4 | 529 | 91 | ||
| AK053685.1 | - | 3 - 4 | 682 | 91 | ||
| BC107254.2 | - | 3 - 4 | 228 | 91 | ||
| BC125012.1 | - | 3 - 4 | 227 | 91 | ||
| BC153828.1 | - | 3 - 4 | 228 | 91 | ||
More Information
Set Membership: |
|

Vstm4
Genomic Map