Assay Details
| Gene Symbol: Vstm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178791.4 | NP_848906.1 | 4 - 5 | 785 | 68 | |
| GenBank mRNA | AK040699.1 | - | 4 - 5 | 633 | 68 | |
| AK053685.1 | - | 4 - 5 | 786 | 68 | ||
| BC107254.2 | - | 4 - 5 | 332 | 68 | ||
| BC125012.1 | - | 4 - 5 | 331 | 68 | ||
| BC153828.1 | - | 4 - 5 | 332 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vstm4
Genomic Map