Assay Details
| Gene Symbol: Dyrk1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113389.1 | NP_001106860.1 | 4 - 5 | 463 | 99 | |
| NM_007890.2 | NP_031916.1 | 4 - 5 | 722 | 99 | ||
| GenBank mRNA | BC129888.1 | - | 3 - 4 | 325 | 40111469 | 99 |
| BC129889.1 | - | 3 - 4 | 325 | 40111470 | 99 | |
| BC138716.1 | - | 3 - 4 | 352 | 8861738 | 99 | |
| BC145405.1 | - | 3 - 4 | 325 | 9053963 | 99 | |
| CB526389.1 | - | 4 - 5 | 463 | 99 | ||
| CD540384.1 | - | 4 - 5 | 438 | 99 | ||
| U58497.1 | - | 3 - 4 | 352 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dyrk1a
Genomic Map