Assay Details
| Gene Symbol: Dyrk1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113389.1 | NP_001106860.1 | 9 - 10 | 1392 | 102 | |
| NM_007890.2 | NP_031916.1 | 9 - 10 | 1651 | 102 | ||
| GenBank mRNA | AK052390.1 | - | 3 - 4 | 527 | 102 | |
| BC034550.1 | - | 4 - 5 | 737 | 102 | ||
| BC129888.1 | - | 8 - 9 | 1254 | 40111469 | 102 | |
| BC129889.1 | - | 8 - 9 | 1254 | 40111470 | 102 | |
| BC138716.1 | - | 8 - 9 | 1281 | 8861738 | 102 | |
| BC145405.1 | - | 8 - 9 | 1254 | 9053963 | 102 | |
| U58497.1 | - | 8 - 9 | 1281 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dyrk1a
Genomic Map