Assay Details
| Gene Symbol: 4932443I19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101519.1 | NP_001094989.1 | 1 - 2 | 171 | 134 | |
| GenBank mRNA | AK038767.1 | - | 1 - 2 | 173 | 134 | |
More Information
Updated Mapping Information
Set Membership: |
|

4932443I19Rik
Genomic Map