Assay Details
| Gene Symbol: Ppp1cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172707.3 | NP_766295.2 | 5 - 6 | 975 | 70 | |
| GenBank mRNA | AK004686.2 | - | 5 - 6 | 686 | 70 | |
| AK088893.1 | - | 5 - 6 | 615 | 70 | ||
| AK089067.1 | - | 794 | 70 | |||
| AK147112.1 | - | 5 - 6 | 778 | 70 | ||
| AK160743.1 | - | 5 - 6 | 684 | 70 | ||
| AK166168.1 | - | 5 - 6 | 893 | 70 | ||
| AK166416.1 | - | 5 - 6 | 979 | 70 | ||
| AK166623.1 | - | 5 - 6 | 878 | 70 | ||
| AK168141.1 | - | 5 - 6 | 872 | 70 | ||
| AK168512.1 | - | 5 - 6 | 621 | 70 | ||
| AK169379.1 | - | 5 - 6 | 724 | 70 | ||
| AK171283.1 | - | 5 - 6 | 844 | 70 | ||
| AK216562.1 | - | 1 - 2 | 35 | 70 | ||
| BC046832.1 | - | 5 - 6 | 676 | 6812937 | 70 | |
| M27073.1 | - | 5 - 6 | 684 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1cb
Genomic Map