Assay Details
| Gene Symbol: Ppp1cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172707.3 | NP_766295.2 | 6 - 7 | 1126 | 136 | |
| GenBank mRNA | AK088893.1 | - | 6 - 7 | 766 | 136 | |
| AK089067.1 | - | 945 | 136 | |||
| AK147112.1 | - | 6 - 7 | 929 | 136 | ||
| AK160743.1 | - | 6 - 7 | 835 | 136 | ||
| AK166168.1 | - | 6 - 7 | 1044 | 136 | ||
| AK168141.1 | - | 6 - 7 | 1023 | 136 | ||
| AK168512.1 | - | 6 - 7 | 773 | 136 | ||
| AK169379.1 | - | 6 - 7 | 875 | 136 | ||
| AK171283.1 | - | 6 - 7 | 995 | 136 | ||
| AK217507.1 | - | 1 - 2 | 162 | 136 | ||
| BC046832.1 | - | 6 - 7 | 827 | 6812937 | 136 | |
| M27073.1 | - | 6 - 7 | 835 | 136 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1cb
Genomic Map