| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500004012.mm.1 |
NM_001177696.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004012.mm.1 |
NM_001177696.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004012.mm.1 |
NM_001177697.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004012.mm.1 |
NM_001177697.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004012.mm.1 |
NM_001177698.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004012.mm.1 |
NM_001177698.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004012.mm.1 |
NM_001177699.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004012.mm.1 |
NM_001177699.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004012.mm.1 |
NM_001177700.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004012.mm.1 |
NM_001177700.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004012.mm.1 |
NM_177678.7.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004012.mm.1 |
NM_177678.7 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004012.mm.1 |
TC0500000400.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500004013.mm.1 |
NM_001177696.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004013.mm.1 |
NM_001177696.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004013.mm.1 |
NM_001177697.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004013.mm.1 |
NM_001177697.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004013.mm.1 |
NM_001177698.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004013.mm.1 |
NM_001177698.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004013.mm.1 |
NM_001177699.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004013.mm.1 |
NM_001177699.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004013.mm.1 |
NM_001177700.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004013.mm.1 |
NM_001177700.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004013.mm.1 |
NM_177678.7.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004013.mm.1 |
NM_177678.7 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004013.mm.1 |
TC0500000400.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500004014.mm.1 |
NM_001177696.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004014.mm.1 |
NM_001177696.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004014.mm.1 |
NM_001177697.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004014.mm.1 |
NM_001177697.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004014.mm.1 |
NM_001177698.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004014.mm.1 |
NM_001177698.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004014.mm.1 |
NM_001177699.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004014.mm.1 |
NM_001177699.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004014.mm.1 |
NM_001177700.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004014.mm.1 |
NM_001177700.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004014.mm.1 |
NM_177678.7.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004014.mm.1 |
NM_177678.7 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004014.mm.1 |
TC0500000400.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500004015.mm.1 |
NM_001177696.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004015.mm.1 |
NM_001177696.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004015.mm.1 |
NM_001177697.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004015.mm.1 |
NM_001177697.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004015.mm.1 |
NM_001177698.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004015.mm.1 |
NM_001177698.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004015.mm.1 |
NM_001177699.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004015.mm.1 |
NM_001177699.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004015.mm.1 |
NM_001177700.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004015.mm.1 |
NM_001177700.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004015.mm.1 |
NM_177678.7.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004015.mm.1 |
NM_177678.7 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004015.mm.1 |
TC0500000400.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500004018.mm.1 |
NM_001177696.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004018.mm.1 |
NM_001177696.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004018.mm.1 |
NM_177678.7.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004018.mm.1 |
NM_177678.7 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004018.mm.1 |
TC0500000400.mm.1 |
MTA TC |