Assay Details
| Gene Symbol: Ablim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177696.1 | NP_001171167.1 | 17 - 18 | 1843 | 77 | |
| NM_177678.7 | NP_808346.3 | 15 - 16 | 1687 | 77 | ||
| GenBank mRNA | AJ748602.1 | - | 15 - 16 | 1643 | 77 | |
| AK046243.1 | - | 15 - 16 | 1638 | 77 | ||
| AK053580.1 | - | 4 - 5 | 373 | 77 | ||
More Information
Set Membership: |
|

Ablim2
Genomic Map