Assay Details
| Gene Symbol: Ifnar2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110498.1 | NP_001103968.1 | 2 - 3 | 384 | 68 | |
| NM_010509.2 | NP_034639.2 | 2 - 3 | 384 | 68 | ||
| GenBank mRNA | AF013274.1 | - | 2 - 3 | 162 | 68 | |
| AF013486.1 | - | 2 - 3 | 256 | 68 | ||
| AK003185.1 | - | 2 - 3 | 318 | 68 | ||
| AK139169.1 | - | 2 - 3 | 364 | 68 | ||
| AK150386.1 | - | 2 - 3 | 318 | 68 | ||
| AK150597.1 | - | 2 - 3 | 297 | 68 | ||
| AK151474.1 | - | 2 - 3 | 312 | 68 | ||
| AK151493.1 | - | 2 - 3 | 297 | 68 | ||
| AK151777.1 | - | 2 - 3 | 312 | 68 | ||
| AK152878.1 | - | 2 - 3 | 318 | 68 | ||
| AK153317.1 | - | 2 - 3 | 318 | 68 | ||
| AK195915.1 | - | 1 - 2 | 103 | 68 | ||
| BC003743.1 | - | 1 - 2 | 79 | 68 | ||
| BC071225.1 | - | 1 - 2 | 82 | 4187603 | 68 | |
| BX515017.1 | - | 2 - 3 | 407 | 68 | ||
| Y09813.1 | - | 2 - 3 | 139 | 68 | ||
| Y09864.1 | - | 2 - 3 | 274 | 68 | ||
| Y09865.1 | - | 2 - 3 | 274 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ifnar2
Genomic Map