Assay Details
| Gene Symbol: Ifnar2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110498.1 | NP_001103968.1 | 6 - 7 | 886 | 142 | |
| NM_010509.2 | NP_034639.2 | 6 - 7 | 886 | 142 | ||
| GenBank mRNA | AF013274.1 | - | 6 - 7 | 664 | 142 | |
| AF013486.1 | - | 6 - 7 | 758 | 142 | ||
| AK003185.1 | - | 6 - 7 | 820 | 142 | ||
| AK010952.1 | - | 6 - 7 | 837 | 142 | ||
| AK139169.1 | - | 6 - 7 | 866 | 142 | ||
| AK150386.1 | - | 6 - 7 | 820 | 142 | ||
| AK150597.1 | - | 6 - 7 | 799 | 142 | ||
| AK151474.1 | - | 6 - 7 | 814 | 142 | ||
| AK151493.1 | - | 6 - 7 | 799 | 142 | ||
| AK151777.1 | - | 6 - 7 | 814 | 142 | ||
| AK152878.1 | - | 6 - 7 | 820 | 142 | ||
| AK153317.1 | - | 6 - 7 | 820 | 142 | ||
| BC003743.1 | - | 5 - 6 | 581 | 142 | ||
| BC071225.1 | - | 5 - 6 | 584 | 4187603 | 142 | |
| Y09813.1 | - | 6 - 7 | 641 | 142 | ||
| Y09864.1 | - | 6 - 7 | 776 | 142 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ifnar2
Genomic Map