Assay Details
| Gene Symbol: Eva1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199210.1 | NP_001186139.1 | 2 - 3 | 769 | 64 | |
| NM_027627.2 | NP_081903.1 | 2 - 3 | 769 | 64 | ||
| GenBank mRNA | AF358257.1 | - | 2 - 3 | 496 | 64 | |
| AK016443.1 | - | 4 - 5 | 765 | 64 | ||
| BC100350.1 | - | 2 - 3 | 515 | 30522196 | 64 | |
| BC132305.1 | - | 2 - 3 | 769 | 40130582 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eva1c
Genomic Map