Assay Details
| Gene Symbol: Hadhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289798.1 | NP_001276727.1 | 2 - 3 | 200 | 114 | |
| GenBank mRNA | AK033462.1 | - | 2 - 3 | 135 | 114 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK033462.1, NM_001289798.1 transcripts.
Set Membership: |
|

Hadhb
Genomic Map