Assay Details
| Gene Symbol: Hadhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289798.1 | NP_001276727.1 | 4 - 5 | 353 | 73 | |
| NM_001289799.1 | NP_001276728.1 | 4 - 5 | 349 | 73 | ||
| NM_145558.2 | NP_663533.1 | 4 - 5 | 306 | 73 | ||
| GenBank mRNA | AK033462.1 | - | 4 - 5 | 288 | 73 | |
| AK076814.1 | - | 4 - 5 | 269 | 73 | ||
| AK083164.1 | - | 4 - 5 | 314 | 73 | ||
| AK083767.1 | - | 4 - 5 | 269 | 73 | ||
| AK150889.1 | - | 4 - 5 | 268 | 73 | ||
| AK168594.1 | - | 4 - 5 | 270 | 73 | ||
| AK169637.1 | - | 4 - 5 | 282 | 73 | ||
| BC005585.1 | - | 3 - 4 | 237 | 3158015 | 73 | |
| BY778233.1 | - | 4 - 5 | 283 | 73 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hadhb
Genomic Map