Assay Details
| Gene Symbol: Hadhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289798.1 | NP_001276727.1 | 5 - 6 | 399 | 152 | |
| NM_001289799.1 | NP_001276728.1 | 5 - 6 | 395 | 152 | ||
| NM_145558.2 | NP_663533.1 | 5 - 6 | 352 | 152 | ||
| GenBank mRNA | AK033462.1 | - | 5 - 6 | 334 | 152 | |
| AK076814.1 | - | 5 - 6 | 315 | 152 | ||
| AK083164.1 | - | 5 - 6 | 360 | 152 | ||
| AK083767.1 | - | 5 - 6 | 315 | 152 | ||
| AK150889.1 | - | 5 - 6 | 314 | 152 | ||
| AK168594.1 | - | 5 - 6 | 316 | 152 | ||
| AK169637.1 | - | 5 - 6 | 328 | 152 | ||
| BC005585.1 | - | 4 - 5 | 283 | 3158015 | 152 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hadhb
Genomic Map