Assay Details
| Gene Symbol: Drc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033460.3 | NP_001028632.1 | 1669 | 92 | ||
| GenBank mRNA | AK140154.1 | - | 1670 | 92 | ||
| BC150863.1 | - | 1667 | 9087774 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Drc1
Genomic Map