Assay Details
| Gene Symbol: Drc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033460.3 | NP_001028632.1 | 2266 | 83 | ||
| GenBank mRNA | AK140154.1 | - | 2267 | 83 | ||
| BC150863.1 | - | 2264 | 9087774 | 83 | ||
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P481279 | NM_001033460.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P481279 | AK140154.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P481279 | NM_001033460.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P481279 | AK140154.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 730766 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 521546 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 139373 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 704179 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 234527 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 940365 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 162013 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 220474 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 763910 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 212713 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 591789 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 562827 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 750303 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 34171 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 400754 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 301915 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 419519 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 292249 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 294447 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 76762 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 23093 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 171153 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 1603 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P481279 | NM_001033460.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P481279 | AK140154.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P481279 | NM_001033460.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P481279 | AK140154.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P103258 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01064386 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01064374 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05519911 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01064398 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05519870 | - | - |
| Whole Mouse Genome G4122A | A_52_P481279 | NM_001033460.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P481279 | AK140154.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P481279 | NM_001033460.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P481279 | AK140154.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P103258 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01064386 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05519870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571818 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04194343 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04771731 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571725 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04153616 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571782 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04460517 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04233879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571777 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571724 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04437740 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04233873 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571738 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571750 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571720 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571754 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571781 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04460515 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04743568 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571721 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571733 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571716 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04806517 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04530391 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04476224 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04437739 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571776 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571723 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04664946 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571735 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571752 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571774 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571718 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571722 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571762 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04233872 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04194344 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04530395 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571711 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04153619 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04778909 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04594774 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04194342 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10520500 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455279_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10520499 | - | - |
| Mouse Genome 430 2.0 Array | 1455279_at | - | - |
| Mouse Genome 430 2.0 Array | 1455279_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 381738_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 381738_PM_at | - | - |
| Mouse Expression Set 430 | 1455279_at | - | - |
| Mouse Expression Set 430 | 1455279_PM_at | - | - |
| Murine Genome U74v2 Set | 162404_i_at | - | - |
| Murine Genome U74v2 Set | 162404_PM_i_at | - | - |
| Murine Genome U74v2 Set | 92377_PM_at | - | - |
| Murine Genome U74v2 Set | 92377_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455279_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455279_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455279_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455279_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1234204 | - | - |

Drc1
Genomic Map