Assay Details
| Gene Symbol: Ube3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133907.3 | NP_598668.1 | 9 - 10 | 1416 | 90 | |
| GenBank mRNA | AK029973.1 | - | 9 - 10 | 1226 | 90 | |
| AK045211.1 | - | 9 - 10 | 1214 | 90 | ||
| AK046056.1 | - | 9 - 10 | 1417 | 90 | ||
| AK122187.1 | - | 9 - 10 | 1351 | 90 | ||
| BC059245.1 | - | 9 - 10 | 1231 | 90 | ||
| BC120731.1 | - | 9 - 10 | 1163 | 40129654 | 90 | |
| BC137626.1 | - | 9 - 10 | 1163 | 8860645 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ube3c
Genomic Map