Assay Details
| Gene Symbol: Whsc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081102.2 | NP_001074571.2 | 1 - 2 | 632 | 89 | |
| NM_001177884.1 | NP_001171355.1 | 1 - 2 | 632 | 89 | ||
| NM_175231.2 | NP_780440.2 | 1 - 2 | 632 | 89 | ||
| GenBank mRNA | BC079668.1 | - | 1 - 2 | 548 | 89 | |
| BC158094.1 | - | 2 - 3 | 706 | 89 | ||
| CF532498.1 | - | 1 - 2 | 169 | 89 | ||
More Information
Set Membership: |
|

Whsc1
Genomic Map