| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500003448.mm.1 |
NM_001081102.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003448.mm.1 |
NM_001081102.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003448.mm.1 |
NM_001177884.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003448.mm.1 |
NM_001177884.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003448.mm.1 |
NM_175231.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003448.mm.1 |
NM_175231.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003448.mm.1 |
TC0500000363.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500003449.mm.1 |
NM_001081102.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003449.mm.1 |
NM_001081102.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003449.mm.1 |
NM_001177884.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003449.mm.1 |
NM_001177884.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003449.mm.1 |
NM_175231.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003449.mm.1 |
NM_175231.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003449.mm.1 |
TC0500000363.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500003451.mm.1 |
NM_001081102.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003451.mm.1 |
NM_001081102.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003451.mm.1 |
NM_001177884.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003451.mm.1 |
NM_001177884.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003451.mm.1 |
NM_175231.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003451.mm.1 |
NM_175231.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003451.mm.1 |
TC0500000363.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500003468.mm.1 |
NM_001081102.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003468.mm.1 |
NM_001081102.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003468.mm.1 |
NM_175231.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003468.mm.1 |
NM_175231.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003468.mm.1 |
TC0500000363.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500003447.mm.1 |
NM_001081102.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003447.mm.1 |
NM_001081102.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003447.mm.1 |
NM_001177884.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003447.mm.1 |
NM_001177884.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003447.mm.1 |
NM_175231.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003447.mm.1 |
NM_175231.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003447.mm.1 |
TC0500000363.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500003452.mm.1 |
NM_001081102.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003452.mm.1 |
NM_001081102.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003452.mm.1 |
NM_001177884.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003452.mm.1 |
NM_001177884.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003452.mm.1 |
NM_175231.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003452.mm.1 |
NM_175231.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003452.mm.1 |
TC0500000363.mm.1 |
MTA TC |