Assay Details
| Gene Symbol: Pls3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166453.1 | NP_001159925.1 | 10 - 11 | 1313 | 118 | |
| NM_001166454.2 | NP_001159926.1 | 10 - 11 | 1308 | 118 | ||
| NM_145629.2 | NP_663604.1 | 10 - 11 | 1427 | 118 | ||
| GenBank mRNA | AB182243.1 | - | 9 - 10 | 1182 | 118 | |
| AK146087.1 | - | 10 - 11 | 1238 | 118 | ||
| AK146455.1 | - | 10 - 11 | 1230 | 118 | ||
| AK160501.1 | - | 10 - 11 | 118 | |||
| AK165540.1 | - | 10 - 11 | 1315 | 118 | ||
| BC005459.1 | - | 10 - 11 | 1388 | 3495462 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pls3
Genomic Map