Assay Details
| Gene Symbol: Pls3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166453.1 | NP_001159925.1 | 4 - 5 | 496 | 108 | |
| NM_001166454.2 | NP_001159926.1 | 4 - 5 | 491 | 108 | ||
| NM_145629.2 | NP_663604.1 | 4 - 5 | 610 | 108 | ||
| GenBank mRNA | AB182243.1 | - | 3 - 4 | 365 | 108 | |
| AK146087.1 | - | 4 - 5 | 421 | 108 | ||
| AK146455.1 | - | 4 - 5 | 413 | 108 | ||
| AK160501.1 | - | 4 - 5 | 439 | 108 | ||
| AK165540.1 | - | 4 - 5 | 496 | 108 | ||
| BC005459.1 | - | 4 - 5 | 571 | 3495462 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pls3
Genomic Map