Assay Details
| Gene Symbol: Mtcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039373.5 | NP_001034462.1 | 3 - 4 | 895 | 83 | |
| GenBank mRNA | U32332.1 | - | 3 - 4 | 854 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mtcp1
Genomic Map