Assay Details
| Gene Symbol: Rngtt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF025653.1 | - | 8 - 9 | 927 | 123 | |
| AF034568.1 | - | 8 - 9 | 981 | 123 | ||
| AK002922.1 | - | 8 - 9 | 1030 | 123 | ||
| AK079488.1 | - | 8 - 9 | 1024 | 123 | ||
| AK142016.1 | - | 8 - 9 | 1024 | 123 | ||
| AK150497.1 | - | 8 - 9 | 1034 | 123 | ||
| AK151463.1 | - | 8 - 9 | 1048 | 123 | ||
| AK160007.1 | - | 8 - 9 | 1036 | 123 | ||
| AK160706.1 | - | 8 - 9 | 1014 | 123 | ||
| AK169305.1 | - | 8 - 9 | 1000 | 123 | ||
| BC043657.1 | - | 8 - 9 | 999 | 5343539 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rngtt
Genomic Map